Learn More
MP Biomedicals™ RNA PLUS
Used for RNA isolation from tissues and cells of human, animal, plant, yeast and bacterial origin
Brand: MP Biomedicals™ 11RNAPLUS2
Code : 50
Additional Details : CAS Number : 108-95-2 Weight : 0.10000kg Transport : UN number : 2810 Chem class : 6.1 Pack group : II
Description
The RNA is suitable for all downstream applications including RT-PCR, hybridization, Northerns, polyA + mRNA isolation, in vitro transcription. RNA-PLUS™ represents an improvement on Chomczynski's procedure for single step RNA isolation providing higher efficiency removal of DNA and proteins. The method promotes formation of complexes of RNA with guanidinium and water molecules and abolishes hydrophilic interactions of DNA and proteins during sample extraction. RNAs obtained with RNA-PLUS™ are not degraded and are free of proteins and DNA. 1mL of reagent is sufficient for 50mg tissue or 5 x 106 cells.
Specifications
Store at 2-8°C. | |
Nucleic Acid Isolation & Purification; Sample Lysis & Preparation | |
RNAPLUS | |
100 mL |
Safety and Handling
- RNA-plus solution
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- Germ cell mutagenicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- Skin sensitiser Category 1
Hazard Statement
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H341-Suspected of causing genetic defects.
- H373-May cause damage to organs.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : phenol, guanidine isothiocyanate