missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Human Albumin
From human plasma
£465.34
Especificaciones
For Use With (Application) | ELISA, Molecular Biology, Western Blot |
---|---|
Molecular Weight (g/mol) | 65 |
Name | Human Albumin |
Source | Human |
Storage Requirements | 4°C |
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
11430412
|
MP Biomedicals™
0219134901 |
1g |
£465.34
1g |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descripción
Negative for HBsAg and HIV antibodies as tested. Purity: >97%. Salt-free, lyophilized, homogeneous powder. MW: 65kDa.
Especificaciones
ELISA, Molecular Biology, Western Blot | |
Human Albumin | |
4°C |
65 | |
Human | |
Lyophilized |