Learn More
MP Biomedicals™ 3,3-Diaminobenzidine Tablets
3,3'-Diaminobenzidine is used in immunohistology and immunoblotting as a precipitating substrate for the localization of peroxidase activity. Insoluble end product is resistant to alcohol and fading, making it ideal for immunohistochemistry. Each tablet contains 5 mg of DAB.
Brand: MP Biomedicals™ 08980681
Code : NEW
Additional Details : CAS Number : 7411-49-6 Weight : 0.10000kg
Description
- Preweighed reagents in tablet form offer convenience and efficiency.
- Tablets both reduce the risk of exposure to hazardous dust and eliminate cumbersome weighing and waste.
- Simply drop into the appropriate amount of distilled water, dissolve, and proceed.
- 100 tablets/Pk.
- Dissolve one tablet in 10 mL deionized water. Use liquid substrate within one hour. Immediately before use, add hydrogen peroxide to 0.1% (v/v).
Specifications
5 mg of DAB per Tablet | |
4.94 | |
Tablet |
Immunohistology, immunoblotting | |
100 x 5 mg |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.