Learn More
Invitrogen™ Mouse LAP/TGFb ProQuantum Immunoassay Kit
The ProQuantum Mouse LAP/TGFbeta Immunoassay Kit is designed to provide quantitative measurements of mouse LAP/TGFbeta in small sample volumes.
£662.00 - £2955.00
Specifications
Accession Number | CED; DPD1; LAP; Latency-associated peptide; prepro-transforming growth factor beta-1; regulatory protein; TGFB; TGFB1; Tgfb-1; TGFbeta; TGF-beta 1; TGFbeta1; TGF-beta1; TGF-beta-1; transforming growth factor beta 1; transforming growth factor beta-1; Transforming growth factor beta-1 proprotein; transforming growth factor, beta 1; transforming growth factor-beta 1 |
---|---|
Assay Range | 0.032 to 2500 ng/mL |
Assay Sensitivity | <0.03 ng/mL |
Product Type | ELISA |
Sample Type | Plasma, Serum, Supernatant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17124303
|
Invitrogen™
A46342 |
96 Tests |
£662.00
Pack of 96 |
Estimated Shipment: 13-02-2025
Log in to see stock. |
|||||
17124749
|
Invitrogen™
A463425 |
5 x 96 Tests |
£2955.00
Each |
Estimated Shipment: 21-02-2025
Log in to see stock. |
|||||
Description
Utilizing proximity-based amplification technology, the assay combines the analyte specificity of high-affinity antibody-antigen binding with the signal detection and amplification capabilities of real-time PCR to achieve a simple yet powerful next-generation protein quantitation platform. A user-friendly workflow combined with intuitive software for analytics enables sample-to-answer in just 2 hours.
- High sensitivity-detect low levels of protein with greater sensitivity than traditional methods
- Broad dynamic range-≥5 logarithmic units, minimizing sample dilutions to ensure they fall within the range
- Small sample consumption-use 2–5 μL of sample (compared to 75 μL for triplicate wells with other methods)
- Fast, easy workflow-2 hours from sample to answer, with no wash steps
- No proprietary instrument to purchase-runs on any real-time PCR instrument
Specifications
CED; DPD1; LAP; Latency-associated peptide; prepro-transforming growth factor beta-1; regulatory protein; TGFB; TGFB1; Tgfb-1; TGFbeta; TGF-beta 1; TGFbeta1; TGF-beta1; TGF-beta-1; transforming growth factor beta 1; transforming growth factor beta-1; Transforming growth factor beta-1 proprotein; transforming growth factor, beta 1; transforming growth factor-beta 1 | |
<0.03 ng/mL | |
Plasma, Serum, Supernatant | |
P04202 | |
Assay Dilution Buffer Antibody-Conjugate A Antibody-Conjugate B Antibody-Conjugate Dilution Buffer Standard Master Mix Ligase | |
Serum, 2-5 μL; Plasma, 2-5 μL; Supernatant, 2-5 μL | |
LAP | |
2 hr |
0.032 to 2500 ng/mL | |
ELISA | |
qPCR Instruments | |
TGFB1 | |
Research Use Only | |
-20°C | |
30 min |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.