missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-5 Mouse Uncoated ELISA Kit with Plates

Specifically engineered for accurate and precise measurement of mouse IL-5 protein levels from samples including serum, lavage fluid, and cell culture supernatants.

Brand:  Invitrogen™ 88-7054-86

 View more versions of this product

Product Code. 15521027

  • £408.00 / Pack of 10
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket

Alert:

Contains Azide


Includes:
  • Capture Antibody: Pre-titrated, purified antibody
  • Detection Antibody. Pre-titrated, biotin-conjugated antibody
  • Standard. Recombinant protein for generating standard curve and calibrating samples
  • 10X Coating Buffer . Buffer for plating the Capture Antibody
  • 5X ELISA/ELISPOT Diluent . Buffer for blocking and diluting the Detection Antibody and Enzyme
  • Enzyme. Pre-titrated Avidin-HRP
  • Substrate. 1X TMB Solution
This item is not returnable. View return policy

Description

Description

The Mouse Interleukin-5 (IL-5) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse IL-5. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

IL-5 is a cytokine that acts as a growth and differentiation factor for both B cells and eosinophils. IL-5 cytokine is a main regulator of eosinopoiesis, eosinophil maturation and activation. The elevated production of this cytokine is reported to be related to asthma or hypereosinophilic syndromes. The receptor of IL-5 cytokine is a heterodimer, whose beta subunit is shared with the receptors for interleukine 3 (IL3) and colony stimulating factor 2 (CSF2/GM-CSF). IL-5, together with those for interleukin 4 (IL4), interleukin 13 (IL13), and CSF2, form a cytokine gene cluster on chromosome 5. IL-5, IL4, and IL13 are found to be regulated coordinately by long-range regulatory elements spread over 120 kilobases on chromosome 5q31.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

P04401
4 pg/mL
ELISA Kit
Mouse
Colorimetric Microplate Reader
GDCF-2,HC11,HSMCR30,MCAF,MCP-1,MCP1,SCYA2,SMC-CF
HRP
RUO
2°C to 8°C
Mouse
24 hr. 30 min.
4-500 pg/mL
Biotin
Lavage Fluid, Serum, Supernatant
ELISA
16191
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; biotin-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Enzyme: Pre-titrated Avidin-HRP, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22; -76; -86)
10 x 96 Tests
Lavage Fluid,100 μL; Serum, 100 μL; Supernatant, 100 μL
Interleukin-5
1 hr. 45 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only