missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOST/Sclerostin Mouse anti-Human, Clone: 220902, R&D Systems™
Mouse Monoclonal Antibody has been used in 7 publications
POA
Specifications
Antigen | SOST/Sclerostin |
---|---|
Immunogen | Mouse myeloma cell line NS0-derived recombinant human SOST/Sclerostin Gln24-Tyr213 Accession # Q9BQB4 |
Host Species | Mouse |
Isotype | IgG2b |
Conjugate | Unconjugated |
Description
SOST/Sclerostin Monoclonal specifically detects SOST/Sclerostin in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
SOST/Sclerostin | |
Mouse | |
Unconjugated | |
RUO |
Mouse myeloma cell line NS0-derived recombinant human SOST/Sclerostin Gln24-Tyr213 Accession # Q9BQB4 | |
IgG2b | |
Western Blot, ELISA | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |