missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERH (M07A), Mouse anti-Human, Clone: 1H4, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant ERH.
Brand: Abnova H00002079-M07A.200uL
Additional Details : Weight : 0.00010kg
Description
Sequence: MSHTILLVQPTKRPEGRTYADYESVNECMEGVCKMYEEHLKRMNPNSPSITYDISQLFDFIDDLADLSCLVYRADTQTYQPYNKDWIKEKIYVLLRRQAQQAGKSpecifications
ERH | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
BC014301 | |
ERH | |
ERH (AAH14301,1 a.a. ∽ 104 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
2079 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
1H4 | |
Mouse monoclonal antibody raised against a full-length recombinant ERH. | |
ERH | |
DROER/FLJ27340 | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |