missing translation for 'onlineSavingsMsg'
Learn More

MOPS sodium salt, 98%, Thermo Scientific Chemicals

£30.85 - £224.12

Chemical Identifiers

CAS 71119-22-7
Molecular Formula C7H14NNaO4S
MDL Number MFCD00064350
InChI Key MWEMXEWFLIDTSJ-UHFFFAOYSA-M
Synonym mops sodium salt, sodium 3-morpholinopropanesulfonate, 4-morpholinepropanesulfonic acid, sodium salt, sodium 3-morpholin-4-ylpropane-1-sulfonate, 3-n-morpholino propanesulfonic acid sodium salt, 3-4-morpholino propanesulfonic acid sodium salt, 4-morpholinepropanesulfonic acid, sodium salt 1:1, 4-morpholinepropanesulfonic acid sodium salt, sodium 3-morpholin-4-yl propane-1-sulfonate, 3-morpholin-4-ylpropanesulfonic acid, sodium salt
PubChem CID 3859613
IUPAC Name sodium;3-morpholin-4-ylpropane-1-sulfonate
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11453437
View Documents
Thermo Scientific Alfa Aesar
A17214.14
25 g
£30.85
25g
Please to purchase this item. Need a web account? Register with us today!
11463437
View Documents
Thermo Scientific Alfa Aesar
A17214.22
100 g
£72.76
100g
Please to purchase this item. Need a web account? Register with us today!
11473437
View Documents
Thermo Scientific Alfa Aesar
A17214.36
500 g
£224.12
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

3-(N-Morpholino)propanesulfonic Acid, Na; an analog to MES used in polyacrylamide gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(N-Morpholino)propanesulfonic Acid, Na; an analog to MES used in polyacrylamide gel electrophoresis.

Solubility
Soluble in water (523 g/L at 20°C).

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
Specifications

Chemical Identifiers

71119-22-7
MFCD00064350
mops sodium salt, sodium 3-morpholinopropanesulfonate, 4-morpholinepropanesulfonic acid, sodium salt, sodium 3-morpholin-4-ylpropane-1-sulfonate, 3-n-morpholino propanesulfonic acid sodium salt, 3-4-morpholino propanesulfonic acid sodium salt, 4-morpholinepropanesulfonic acid, sodium salt 1:1, 4-morpholinepropanesulfonic acid sodium salt, sodium 3-morpholin-4-yl propane-1-sulfonate, 3-morpholin-4-ylpropanesulfonic acid, sodium salt
sodium;3-morpholin-4-ylpropane-1-sulfonate
C7H14NNaO4S
MWEMXEWFLIDTSJ-UHFFFAOYSA-M
3859613

Specifications

∽260°C (decomposition)
98%
MFCD00064350
Soluble in water (523g/L at 20°C).
C1COCCN1CCCS(=O)(=O)[O-].[Na+]
231.242
231.25
71119-22-7
C7H14NNaO4S
mops sodium salt, sodium 3-morpholinopropanesulfonate, 4-morpholinepropanesulfonic acid, sodium salt, sodium 3-morpholin-4-ylpropane-1-sulfonate, 3-n-morpholino propanesulfonic acid sodium salt, 3-4-morpholino propanesulfonic acid sodium salt, 4-morpholinepropanesulfonic acid, sodium salt 1:1, 4-morpholinepropanesulfonic acid sodium salt, sodium 3-morpholin-4-yl propane-1-sulfonate, 3-morpholin-4-ylpropanesulfonic acid, sodium salt
MWEMXEWFLIDTSJ-UHFFFAOYSA-M
sodium;3-morpholin-4-ylpropane-1-sulfonate
3859613
MOPS sodium salt
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only