missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOPS, 101%, MP Biomedicals™
Click to view available options
Quantity:
250 g
Specifications
Color | White |
Physical Form | Crystalline Powder |
Quantity | 250 g |
Assay Percent Range | 1.01 |
Formula Weight | 231.2 |
Chemical Name or Material | MOPS |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'