missing translation for 'onlineSavingsMsg'
Learn More

Mono-Ethyl malonate, 95%, Thermo Scientific Chemicals

£28.14 - £688.43

Chemical Identifiers

CAS 1071-46-1
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.11
MDL Number MFCD00020490
InChI Key HGINADPHJQTSKN-UHFFFAOYSA-N
Synonym ethyl hydrogen malonate, monoethyl malonate, monoethyl malonic acid, mono-ethyl malonate, monoethyl hydrogen malonate, malonic acid monoethyl ester, ethoxycarbonyl acetic acid, 2-ethoxycarbonyl acetic acid, 3-ethoxy-3-oxo-propanoic acid, ethylhydrogenmalonate
PubChem CID 70615
ChEBI CHEBI:86907
IUPAC Name 3-ethoxy-3-oxopropanoic acid
SMILES CCOC(=O)CC(=O)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10673205
View Documents
Thermo Scientific Acros
399900050
5 g Glass bottle
£28.14
5g
Please to purchase this item. Need a web account? Register with us today!
10443274
View Documents Promotion Details
Thermo Scientific Acros
399900250
25 g Glass bottle
£83.92
25g
Please to purchase this item. Need a web account? Register with us today!
13448548
View Documents
Thermo Scientific Acros
399902500
250 g Glass bottle
£688.43
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1071-46-1
132.11
HGINADPHJQTSKN-UHFFFAOYSA-N
70615
3-ethoxy-3-oxopropanoic acid
C5H8O4
MFCD00020490
ethyl hydrogen malonate, monoethyl malonate, monoethyl malonic acid, mono-ethyl malonate, monoethyl hydrogen malonate, malonic acid monoethyl ester, ethoxycarbonyl acetic acid, 2-ethoxycarbonyl acetic acid, 3-ethoxy-3-oxo-propanoic acid, ethylhydrogenmalonate
CHEBI:86907
CCOC(=O)CC(=O)O

Specifications

1071-46-1
100.0
106.5°C (3.0 mmHg)
C5H8O4
MFCD00020490
1.119
Solubility in water: soluble. Other solubilities: soluble in other solvents
CCOC(=O)CC(=O)O
132.11
CHEBI:86907
95%
94.0
1.1190g/mL
95%
C2H5O2CCH2CO2H
02,IV,188
ethyl hydrogen malonate, monoethyl malonate, monoethyl malonic acid, mono-ethyl malonate, monoethyl hydrogen malonate, malonic acid monoethyl ester, ethoxycarbonyl acetic acid, 2-ethoxycarbonyl acetic acid, 3-ethoxy-3-oxo-propanoic acid, ethylhydrogenmalonate
HGINADPHJQTSKN-UHFFFAOYSA-N
3-ethoxy-3-oxopropanoic acid
70615
132.11
Mono-Ethyl malonate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 213-992-7

Product Identifier
  • mono-Ethyl malonate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only