missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum, AAS standard solution, Specpure™ Mo 1000μg/mL, Thermo Scientific Chemicals

£30.55 - £63.32

Chemical Identifiers

MDL Number MFCD00003465
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11338657
View Documents
Thermo Scientific Alfa Aesar
035764.AE
100 mL
£30.55
100mL
Please to purchase this item. Need a web account? Register with us today!
11405070
View Documents
Thermo Scientific Alfa Aesar
035764.AP
500 mL
£63.32
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

MFCD00003465

Specifications

Mo in 5% HNO3/tr. HF
UN3264
Liquid
MFCD00003465
Specpure™
Molybdenum AAS standard solution, Mo 1000μg/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H314-H318-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 231-714-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.