missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phen Green™ SK, Dipotassium Salt
Green-fluorescent heavy metal indicator can be used to detect broad range of ions
Brand: Invitrogen™ P14312
This item is not returnable.
View return policy
Description
- Detects Cu2+, Cu+, Fe22+, Hg2+, Pb2+, Cd2+, Zn2+ and Ni2+
Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling, Zinc and Heavy Metal Ion Detection
Order Info
Shipping Condition: Room temperature

Specifications
Store at room temperature and protect from light. | |
Potassium Assay | |
SK Dipotassium Salt | |
Phen Green™ | |
Room Temperature |
Fluorescence | |
Fluorescence Microscope, Microplate Reader | |
Zinc Indicator, Iron Indicator, Nickel Indicator, Mercury Indicator, Cadmium Indicator, Copper Indicator, Lead Indicator | |
1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction