Learn More
Invitrogen™ Molecular Probes™ SlowFade™ Antifade Kit
Protects dyes from fading (photobleaching) during fluorescence microscopy experiments
Brand: Invitrogen™ S2828
Code : 50
Additional Details : Weight : 0.16800kg
Description
- Contains two formulations of the SlowFade™ Antifade Reagent: an aqueous formulation in buffer and an aqueous formulation that contains 50% glycerol
- Both formulations reduce the photobleaching rate of fluorescein to almost zero and are useful for quantitative measurements
- Glycerol-based mountant is ideal for immediate sample viewing
- Contains oxygen scavenger 1,4-diaza bicyclo (2.2.2) octane (DABCO)
- Improved Signal Stability for Fluorescein
- Can extend the useful fluorescence emission of fluorescein more than 50-fold and can preserve the signal in cell and tissue mounts for up to two years
- Substantially quenches the initial fluorescence of fluorescein and almost completely quenches that of the Alexa Fluor 350, Alexa Fluor 405, and Cascade Blue™ fluorophores
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Counterstaining, Mounting and Fade Prevention
Order Info
Shipping Condition: Room Temperature
Specifications
Store at Room temperature. | |
1 kit |
For Research Use Only. Not for use in diagnostic procedures.