missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Histone H1 From Calf Thymus, Alexa Fluor™ 488 Conjugate
Labeled with the bright and photostable dye
Brand: Invitrogen™ H13188
157.74 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
- Exhibits fluorescence with excitation/emission maxima approx. 495/520nm and can be visualized by fluorescence microscopy or flow cytometry
Cell Analysis, Cell Cycle, Cell Structure, Cell Viability, Proliferation and Function, Endocytosis, Exocytosis and Phagocytosis, Lysosomes, Vacuoles
Order Info
Shipping Condition: Room Temperature
Specifications
Store in freezer (-5 to -30°C) and protect from light. | |
Lipopolysaccharide Conjugate | |
Alexa Fluor™ | |
Phosphate Buffered Saline | |
Histone H1 |
Fluorescence | |
Alexa Fluor Dyes | |
1 mg | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.