Learn More
Invitrogen™ Molecular Probes™ EnzChek™ Peptidase/Protease Assay Kit
Quantify the amount of enzyme activity
Brand: Invitrogen™ E33758
Description
The EnzChek Peptidase/Protease Assay Kit enables simple and accurate quantification of a wide range of protease activities within 60 minutes. The EnzChek peptidase/protease substrate included in the kit has been labeled with a fluorophore and a quencher moiety separated by a peptide. Upon cleavage by protease(s), the fluorophore separates from the quencher and, upon excitation, the fluorophore emits fluorescence signal. The magnitude of the resultant signal is proportional to the degree of substrate cleavage, resulting in an ability to quantify the amount of enzyme activity. The assay is performed in a simple mix–incubate–read format, can be completed in 60 minutes or less, and is easily adapted to accommodate diverse pH requirements.
- Simple mix-incubate-read format
- Excitation and emission wavelengths (502/528nm) well suited for fluorescein (FITC) filter settings
- Easily adapted to a variety of proteases and reaction conditions
Cell Analysis, Cellular Imaging, Enzyme and Protein Activity Assays, High-Content Screening (HCS), Protease/Peptidase Activity, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room temperature

Specifications
1 kit | |
Kit, Assay | |
Peptidases, Proteases | |
Store in freezer (-5°C to -30°C) and protect from light. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.