Learn More
Epredia™ Modified Grocott' s Methenamine Silver (Periodic Acid) Stain Kit
Distinguish fungi, basement membranes and opportunistic organisms such as Pneumocystis Carinii in tissue sections with Epredia™ Modified Grocott's Methenamine Silver (Periodic Acid)
Brand:  Epredia™ 88026
 View more versions of this product
Product Code. 12583647
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Fungi, basement membranes and opportunistic organisms, along with reticular fibers, are stained black
  • Background is stained green
Specifications
Specifications
Gold Chloride Solution (0.1%)
125mL