Learn More
Miele Professional™ Miele Process Documentation
Printer PRT/1 built-in for process documentation (G 7824)
Brand:  Miele Professional™ 6224720
Product Code. 15430485
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • 6-needle printer
  • Paper size 58 mm wide
  • Can be installed on the operator side
Specifications
Specifications
Process/Operating Documentation/Communication
Washer-disinfectors, laboratory