Learn More
Fisherbrand™ MicroSon Cleaning Solution
Provides cleaning and decontamination for ultrasonic baths and splash-cleaning. Fisherbrand™ MicroSon Cleaning Solution removes grease, oil, glue, dust and fingerprints.
Brand: Fisherbrand™
Code : A0
Additional Details : CAS Number : 69011-36-5 Weight : 1.04220kg Transport : UN number : 1719 Chem class : 8 Pack group : III
Description
- Green in color
- For glassware, stainless steel and ferrous metals
Specifications
Liquid | |
Dark green | |
2% |
11.3 | |
≥100°C | |
1 L |
Safety and Handling
- MicroSon
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
Hazard Statement
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H301-Toxic if swallowed.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : isotridecanol ethoxylated, C10 fatty alcohol ethoxylated, potassium hydroxide, tetrapotassium pyroph