Learn More
Greiner Bio-One Microlon™ Polystyrene 96-well ELISA Microplates
Manufactured from crystal clear polystyrene for optimum visibility. Greiner BioOne™ Microlon™ Polystyrene 96-Well ELISA Microplates are tested in a colorimetric immunoasay for immunological research applications.
£112.00
Specifications
Format Standard
Lid Without Lid
Array 8 x 12
Color Clear
Material Polystyrene
View More Specs
Products 4
Product Code Brand Binding Property Surface Treatment Well Design Price Quantity & Availability  
Product Code Brand Binding Property Surface Treatment Well Design Price Quantity & Availability  
10577091
View Documents
Greiner Bio-One
655061
High Binding Microlon 600 Standard Well
£112.00
Pack of 40
10144771
View Documents
Greiner Bio-One
655001
Medium Binding Microlon 200 Standard Well
£112.00
Pack of 40
10043211
View Documents
Greiner Bio-One
655081
Medium Binding Microlon 600 Chimney Well
£112.00
Pack of 40
10680932
View Documents
Greiner Bio-One
655080
Medium Binding Microlon 200 Chimney Well
£112.00
Pack of 40
Description
Description
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Tracebility from production through a defined LOT number system
  • Manufactured from crystal clear polystyrene
  • The footprint is compatible with automated systems
  • Well to well spacing : 9 mm
  • Alphanumeric well coding
  • Suitable for transmission measurements
  • Specifically treated to provide an incresed protein binding
  • Well profile: F-bottom / standard (flat bottom) 
Specifications
Specifications
Standard
8 x 12
Polystyrene
Flat Bottom (F-well)
Without Lid
Clear
96
40/Cs.