Learn More
Ansell Microgard™ 2500 Standard Coveralls with Hood
Product Code. 15516574
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Achieves the highest level of protection for barrier to infective agents. Ansell Microgard 2500 Standard Coveralls with Hood offer exceptional mechanical strength, liquid and particulate protection.
  • Category III Coverall with 3-piece hood
  • Ultrasonically welded seams
  • Elasticated wrists, finger loops, waist and ankles.
  • Front zipper and re-sealable storm flap.
  • Protects from biological agents for penetration of blood, body fluids and blood-borne pathogens
  • Moisture vapor permeable to help reduce the risk of heat stress
  • Anti-static according to EN 1149-5
  • Low linting - Reduced risk of contamination in critical areas
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Coveralls
Size 2X-Large
Gender Unisex
Material Microporous Polypropylene Laminate
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Thumb Loop
Neck Style Hooded
Hood Type 3-panel
Closure Type Front Zipper with Storm Flap
Seam Type Welded
Sterility Non-sterile
Certifications/Compliance (Europe) Cat. III Type 4-B, 5-B, 6-B, EN1073-2, EN14126, EN1149-1
Model WH25-W-00-111
No. of Pockets None
Style Includes Hood, Fingerloops