Learn More
Ansell Microgard™ 2300 Comfort Coverallls with Hood
Product Code. 15516584
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
MICROGARD™ 2300 Comfort Coveralls with Hood have been engineered with workers involved in composites manufacturing and related industries in mind, to offer protection where you need it most and ventilation to help reduce the risk of heat stress.
  • Category III Coverall with 3-piece hood ensures a good fit with half and full face respirators
  • Elasticated wrists, waist and ankles.
  • Elasticated finger loops prevent sleeve movement whilst working above your head Bound yellow seams
  • 2-way front zipper with re-sealable storm flap.
  • Breathable SMS back panel.
  • Hood, arms, legs and front torso in MICROGARD™ 2300 fabric
  • Ensures that there is no risk of delamination should a wearer make contact with a tacky surface
  • Air and water vapor permeable back panel ventilates the suit, to help reduce the risk of heat stress
  • Silicone-free for spray painting applications
  • Anti-static certified in accordance with EN 1149-5
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Coveralls
Size X-Large
Gender Unisex
Material Spunbond Polypropylene
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic, Finger Loops
Neck Style Hooded
Hood Type 3-panel, Elastic
Closure Type Front Zipper with Storm Flap
Seam Type Bound
Sterility Non-sterile
Back Style Breathable Microgard™ 1500 Plus
Certifications/Compliance (Europe) EN ISO 9237, EN 1149-5, EN 1073-2
Model WY23-B-00-129
No. of Pockets None
Style Includes Hood, Fingerloops