missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek MultiFlo FX/EL406 Dispenser Cassettes
GREENER_CHOICE

Dispense Cassettes for use with MultiFlo FX Dispenser and EL406 Washer Dispenser.

Brand:  Agilent BioTek 7170015

 View more versions of this product

Product Code. 12531547

  • £1555.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Volume range 1 μL to 10 mL
  • Accommodates 6- to 1536-well microplates, PCR trays, deep-well microplates, microtubes and other tube configurations up to 4 in. in height
  • Dispense 1536 wells in 18 s.
  • Cassettes can be autoclaved without recalibration
  • Dispense up to eight reagents at one time
  • Customizable dispense pattern allows skipped columns or rows
  • Built-in priming trough
  • Easy maintenance of individually replaceable tips and choice of replacement cassettes or tubing
  • Compatible with Bio-Stack microplate stacker 11-120-601 for increased throughput

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 to 50 μL
0.019 cm
1 μL
Low Volume Cassette
0.0075 in.
For liquid handling
Stainless Steel
EL406 Microplate Washer, Multiflo Dual Syringe Dispenser Module, Agilent BioTek MultiFlo FX Multimode Dispenser
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.