Learn More
Applied Biosystems™ MicroAmp™ EnduraPlate™ Optical 96-Well Fast Multicolor Reaction Plates with Barcode
Ideal solution for work that requires special handling and an even greater degree of durability for use with multi-instrument experiments
£28.14 - £2690.00
Specifications
Barcode | Includes Barcode |
---|---|
Certifications/Compliance | Certified DNA/RNase Free |
Content And Storage | Store at room temperature. |
Product Line | MicroAmp |
Color | Optical, Assorted |
Description
Use of polypropylene and a high temperature molding process helps prevent DNA, RNA, and enzymes from binding, while polycarbonate provides mechanical stability, flatness, and excellent durability. ANSI/SBS compliant construction combined with mechanical durability during and after thermal cycling makes EnduraPlate reaction plates an ideal choice for use with automation/robotics and multi-instrument experiments (e.g., thermal cycler, real-time thermal cycler, genetic analyzer).
- Validated with Applied Biosystems instruments for optimal fit and performance
- Certified DNA-, RNAse-, and PCR inhibitor-free with in-process sampling tests
- Constructed to ANSI/SBS standard, well suited to high-throughput laboratories and robotic handling
- Thin-wall construction allows for a snug fit to thermal blocks with excellent heat transfer from thermal block to sample
- Easy-to-read, black well identification text helps to minimize pipetting mistakes and improves sample preparation productivity
- Available in multiple colors for easy organization of laboratory assays
- Excellent mechanical stability and flatness for all workflow needs
- GPLE (For Laboratory Use) version is also available
- Platform: 310 Genetic Analyzer, 3130 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Dx Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer, 7500 Fast Dx System, 7500 Fast System, 7900HT Fast System, StepOnePlus™, Veriti™ Dx Fast Thermal Cycler, Veriti Fast Thermal Cycler, ViiA™ 7 Dx Fast System, ViiA 7 Fast System
PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)
Specifications
Includes Barcode | |
Store at room temperature. | |
Optical, Assorted | |
Thermal Cycler, Genetic Analyzer, DNA Analyzer | |
Polycarbonate, Polypropylene | |
96 | |
Fast |
Certified DNA/RNase Free | |
MicroAmp | |
Microplate | |
Semi | |
0.2 mL | |
0.1 mL |