missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius MGA Grade Glass Microfiber Filters w/o Binder
Binder free glass microfiber filters are suitable for analytical and gravimetric analyses. They are also used as prefilter.
Brand: Sartorius FT-3-1101-150
Code : NEW
Additional Details : Weight : 0.10000kg
Description
Applications include: Clarification of buffer & reagent solutions, Filtration of algae cells.Specifications
36mbar | |
Disc | |
1.6 μm | |
40 sec. | |
MGA | |
500°C | |
0.25 mm | |
15 cm |
White | |
Borosilicate Glass Microfiber | |
Non-sterile | |
Clarification of buffer and reagent solutions | |
932°F | |
0.009 in. | |
5.9 in. |