missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ Density Kits
For use with XS/XP balances. Mettler Toledo Density Kits help to determine the density of solids and liquids with top-loading electronic balances.
£1309.10
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
10567945
|
Mettler Toledo™
11106706 |
Solids kit includes 200mL beaker, thermometer, bridge, sample holder (2), bracket, bottle of wetting agent (25mL) |
£1309.10
1 set |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Solids Kit
- 200mL beaker
- Thermometer
- Bridge
- Sample Holder (2)
- Bracket
- Bottle of wetting agent, 25mL
- Sinker mounted with pan and platinum wire
Note
Both kits are required to perform liquid density measurements.
Specifications
XS/XP Analytical Balance |