Learn More
METTL21B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560204
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TFPLLLGTLQ HLCRPHGTIY LASKMRKEHG TESFFQHLLP QHFQLELAQR DEDENVNIYR ARHREPRPA Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 83%.
METTL21B Polyclonal antibody specifically detects METTL21B in Human samples. It is validated for Immunohistochemistry.Specifications
METTL21B | |
Polyclonal | |
Unconjugated | |
EEF1AKMT3 | |
EEF1A lysine methyltransferase 3; EEF1AKMT3; FAM119B; family with sequence similarity 119, member B; HCA557A; Hepatocellular carcinoma-associated antigen 557a; hepatocellularcarcinoma-associated antigen HCA557a; methyltransferase like 21B; methyltransferase-like protein 21B; METTL21B; Protein-lysine methyltransferase METTL21B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25895 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96AZ1 | |
EEF1AKMT3 | |
Recombinant protein corresponding to Human METTL21B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only