Learn More
METTL13 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562828
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LALLVVGLGG GSLPLFVHDH FPKSCIDAVE IDPSMLEVAT QWFGFSQSDR MKVHIADGLD YIASLAGGGE ARPCYDVIMF DVDSKDPTLG MSCP Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 27%.
METTL13 gene ontology annotations related to this gene include methyltransferase activity.Specifications
METTL13 | |
Polyclonal | |
Unconjugated | |
EEF1AKNMT | |
5630401D24Rik; antiapoptotic protein FEAT; CGI-01; eEF1A lysine and N-terminal methyltransferase; eEF1A lysine methyltransferase; eEF1A N-terminal methyltransferase; EEF1AKNMT; eEF1A-KNMT; feat; KIAA0859; methyltransferase like 13; methyltransferase-like protein 13; METTL13; RGD1311526 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51603 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N6R0 | |
EEF1AKNMT | |
Recombinant protein corresponding to Human METTL13 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only