missing translation for 'onlineSavingsMsg'
Learn More

Methyltrichlorosilane, 98+%, Thermo Scientific Chemicals

£22.61 - £107.74

Chemical Identifiers

CAS 75-79-6
Molecular Formula CH3Cl3Si
Molecular Weight (g/mol) 149.48
InChI Key JLUFWMXJHAVVNN-UHFFFAOYSA-N
Synonym methyltrichlorosilane, trichloro methyl silane, silane, trichloromethyl, methyl-trichlorsilan, methylsilicochloroform, trichloromethylsilicon, methylsilyl trichloride, monomethyltrichlorosilane, silane, methyltrichloro, trichlor-methylsilan
PubChem CID 6399
IUPAC Name trichloro(methyl)silane
SMILES C[Si](Cl)(Cl)Cl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10606332
SDS View Documents
Thermo Scientific Acros
127930100
10 mL Glass bottle
£22.61
10mL
Please to purchase this item. Need a web account? Register with us today!
10705051
SDS View Documents
Thermo Scientific Acros
127935000
500 mL Glass bottle
£40.30
500mL
Please to purchase this item. Need a web account? Register with us today!
11905801
SDS View Documents
Thermo Scientific Acros
127930025
2.5 L Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

75-79-6
149.48
methyltrichlorosilane, trichloro methyl silane, silane, trichloromethyl, methyl-trichlorsilan, methylsilicochloroform, trichloromethylsilicon, methylsilyl trichloride, monomethyltrichlorosilane, silane, methyltrichloro, trichlor-methylsilan
trichloro(methyl)silane
CH3Cl3Si
JLUFWMXJHAVVNN-UHFFFAOYSA-N
6399
C[Si](Cl)(Cl)Cl

Specifications

75-79-6
Colorless
66.0°C
Authentic
CH3Cl3Si
1.4100 to 1.4120
1.27
Solubility in water: reacts
C[Si](Cl)(Cl)Cl
149.48
0.48 mPa.s (20°C)
98+%
Methyltrichlorosilane
-77.0°C
1.2700g/mL
−15°C
98% min. (GC)
CH3SiCl3
11,553; 12,527
methyltrichlorosilane, trichloro methyl silane, silane, trichloromethyl, methyl-trichlorsilan, methylsilicochloroform, trichloromethylsilicon, methylsilyl trichloride, monomethyltrichlorosilane, silane, methyltrichloro, trichlor-methylsilan
JLUFWMXJHAVVNN-UHFFFAOYSA-N
trichloro(methyl)silane
6399
149.48
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Toxic if inhaled.
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Reacts violently with water.
Corrosive to the res

GHS P Statement
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOW

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 200-902-6

Product Identifier
  • Methyltrichlorosilane

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH071-Corrosive to the respiratory tract.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.