missing translation for 'onlineSavingsMsg'
Learn More

Methyle3-hydroxybutanoate, 97%, Thermo Scientific™

Product Code. 10526334
Click to view available options
Quantity:
10 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 3976-69-0
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00063289
InChI Key LDLDJEAVRNAEBW-SCSAIBSYSA-N
Synonym methyl r---3-hydroxybutyrate, r-methyl 3-hydroxybutanoate, methyl 3r-3-hydroxybutanoate, methyl r-3-hydroxybutyrate, r-3-hydroxybutyric acid methyl ester, methyl r-3-hydroxybutanoate, r---3-hydroxybutyric acid methyl ester, r---methyl 3-hydroxybutyrate, r---3-hydroxy-n-butyric acid methyl ester
PubChem CID 2724279
IUPAC Name methyl (3R)-3-hydroxybutanoate
SMILES CC(CC(=O)OC)O

Specifications

CAS Min % 97.0
Density 1.0500g/mL
Assay Percent Range 97%
Linear Formula CH3CH(OH)CH2CO2CH3
Quantity 25 g
Specific Rotation Condition - 21.50 (20.00°C neat)
Formula Weight 118.13
Percent Purity 97%
Chemical Name or Material Methyl 3-hydroxybutanoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'