Learn More
Thermo Scientific Chemicals Methyl Red Sodium Salt, Reagent ACS
£18.19 - £52.36
Chemical Identifiers
CAS | 845-10-3 |
---|---|
Molecular Formula | C15H14N3NaO2 |
Molecular Weight (g/mol) | 291.286 |
MDL Number | MFCD00002426 |
InChI Key | GNTPCYMJCJNRQB-UHFFFAOYSA-M |
Synonym | 2-[4-(Dimethylamino)phenylazo]benzoic acid, sodium salt, C.I. 13020, MR |
PubChem CID | 4465632 |
IUPAC Name | sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate |
SMILES | CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10065700
|
Thermo Scientific Chemicals
414840100 |
10 g | Glass bottle |
£18.19
10g |
Estimated Shipment: 31-01-2025
Log in to see stock. |
||||
10371491
|
Thermo Scientific Chemicals
414840250 |
25 g | Glass bottle |
£23.32
25g |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
10509983
|
Thermo Scientific Chemicals
414841000 |
100 g | Glass bottle |
£52.36
100g |
Estimated Shipment: 13-02-2025
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
845-10-3 | |
291.286 | |
GNTPCYMJCJNRQB-UHFFFAOYSA-M | |
4465632 | |
CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+] |
C15H14N3NaO2 | |
MFCD00002426 | |
2-[4-(Dimethylamino)phenylazo]benzoic acid, sodium salt, C.I. 13020, MR | |
sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate |
Specifications
845-10-3 | |
C15H14N3NaO2 | |
MFCD00002426 | |
15, 6192 | |
GNTPCYMJCJNRQB-UHFFFAOYSA-M | |
CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+] | |
291.286 | |
291.27 | |
From pH 4.2 (pink) to pH 6.2 (yellow) | |
Orange/Red to Yellow | |
Methyl Red, sodium salt, Pure |
(Aqueous) Passes Test | |
(CH3)2NC6H4N=NC6H4CO2Na | |
16, 329 | |
2-[4-(Dimethylamino)phenylazo]benzoic acid, sodium salt, C.I. 13020, MR | |
Authentic | |
sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate | |
4465632 | |
ACS Reagent | |
13020 | |
Liquid |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 212-682-9
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.