missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Orange solution, Honeywell Fluka™
Indicator, 0.1%, Reag. Ph. Eur.
£54.78 - £63.51
Chemical Identifiers
CAS | 547-58-0 |
---|---|
Molecular Formula | C14H14N3NaO3S |
Molecular Weight (g/mol) | 327.334 |
MDL Number | MFCD00007502 |
InChI Key | STZCRXQWRGQSJD-UHFFFAOYSA-M |
PubChem CID | 23673835 |
IUPAC Name | sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate |
SMILES | CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
547-58-0 | |
327.334 | |
STZCRXQWRGQSJD-UHFFFAOYSA-M | |
sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate |
C14H14N3NaO3S | |
MFCD00007502 | |
23673835 | |
CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+] |
Specifications
547-58-0 | |
MFCD00007502 | |
4732884 | |
CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+] | |
327.334 | |
327.33g/mol | |
39°C (102.2°F) |
C14H14N3NaO3S | |
UN1993 | |
STZCRXQWRGQSJD-UHFFFAOYSA-M | |
sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate | |
23673835 | |
0.10% | |
Methyl Orange solution |
Safety and Handling
P210-P403 + P235
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Methyl Orange solution
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : Sodium 4-(4-dimethylaminophenylazo)benzenesulphonate
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title