Learn More
Methyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate, 95%, Thermo Scientific Chemicals
£56.76 - £176.00
Chemical Identifiers
CAS | 342613-63-2 |
---|---|
Molecular Formula | C9H7BrN2O2 |
Molecular Weight (g/mol) | 255.071 |
MDL Number | MFCD10566741 |
InChI Key | VFFFBWRUZMAHFS-UHFFFAOYSA-N |
Synonym | methyl 3-bromoimidazo 1,2-a pyridine-7-carboxylate, 3-bromo-7-methoxycarbonyl imidazo 1,2-a pyridine, methyl 3-bromoh-imidazo 1,2-a pyridine-7-carboxylate, 3-bromoimidazo 1,2-a pyridine-7-carboxylic acid methyl ester, pubchem23890, methyl3-bromoimidazo 1,2-a pyridine-7-carboxylate, 3-bromo-imidazo 1,2-a pyridine-7-carboxylic acid methyl ester, methyl 3-bromo-5h-imidazo 2,1-b 1,3 oxazine-7-carboxylate, 3-bromo-imidazo 1,2-alpha pyridine-7-carboxylic acid methyl ester, 3-bromoimidazo 1,2-alpha pyridine-7-carboxylic acid methyl ester |
PubChem CID | 45790121 |
IUPAC Name | methyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate |
SMILES | COC(=O)C1=CC2=NC=C(N2C=C1)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15439065
|
Thermo Scientific Alfa Aesar
H35994.MD |
250 mg | N/A | ||||||
15429065
|
Thermo Scientific Alfa Aesar
H35994.03 |
1 g |
£176.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Methyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate is used as pharmaceutical intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
342613-63-2 | |
255.071 | |
VFFFBWRUZMAHFS-UHFFFAOYSA-N | |
45790121 | |
COC(=O)C1=CC2=NC=C(N2C=C1)Br |
C9H7BrN2O2 | |
MFCD10566741 | |
methyl 3-bromoimidazo 1,2-a pyridine-7-carboxylate, 3-bromo-7-methoxycarbonyl imidazo 1,2-a pyridine, methyl 3-bromoh-imidazo 1,2-a pyridine-7-carboxylate, 3-bromoimidazo 1,2-a pyridine-7-carboxylic acid methyl ester, pubchem23890, methyl3-bromoimidazo 1,2-a pyridine-7-carboxylate, 3-bromo-imidazo 1,2-a pyridine-7-carboxylic acid methyl ester, methyl 3-bromo-5h-imidazo 2,1-b 1,3 oxazine-7-carboxylate, 3-bromo-imidazo 1,2-alpha pyridine-7-carboxylic acid methyl ester, 3-bromoimidazo 1,2-alpha pyridine-7-carboxylic acid methyl ester | |
methyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate |
Specifications
342613-63-2 | |
C9H7BrN2O2 | |
methyl 3-bromoimidazo 1,2-a pyridine-7-carboxylate, 3-bromo-7-methoxycarbonyl imidazo 1,2-a pyridine, methyl 3-bromoh-imidazo 1,2-a pyridine-7-carboxylate, 3-bromoimidazo 1,2-a pyridine-7-carboxylic acid methyl ester, pubchem23890, methyl3-bromoimidazo 1,2-a pyridine-7-carboxylate, 3-bromo-imidazo 1,2-a pyridine-7-carboxylic acid methyl ester, methyl 3-bromo-5h-imidazo 2,1-b 1,3 oxazine-7-carboxylate, 3-bromo-imidazo 1,2-alpha pyridine-7-carboxylic acid methyl ester, 3-bromoimidazo 1,2-alpha pyridine-7-carboxylic acid methyl ester | |
COC(=O)C1=CC2=NC=C(N2C=C1)Br | |
255.071 | |
255.07 | |
Methyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate |
156°C to 157°C | |
MFCD10566741 | |
VFFFBWRUZMAHFS-UHFFFAOYSA-N | |
methyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate | |
45790121 | |
95% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only