missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 2-ethylacetoacetate, 97%

£25.32 - £96.04

Chemical Identifiers

CAS 51756-08-2
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD01632136
InChI Key YXLVLOWNJCOOAU-UHFFFAOYSA-N
Synonym methyl 2-ethylacetoacetate, butanoic acid, 2-ethyl-3-oxo-, methyl ester, methyl-2-ethylacetoacetate, 2-ethyl-3-oxobutanoic acid, methyl ester, methyl ethylacetoacetate, methyl acetylbutyrate, methyl 2-acetylbutyrate, acmc-1b06q, methyl 2-ethyl-3-oxobutyrate, methyl 2-ethyl-3-oxo-butyrate
PubChem CID 103534
IUPAC Name methyl 2-ethyl-3-oxobutanoate
SMILES CCC(C(=O)C)C(=O)OC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11489307
View Documents
Alfa Aesar™
B21368.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11499307
View Documents
Alfa Aesar™
B21368.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

51756-08-2
144.17
YXLVLOWNJCOOAU-UHFFFAOYSA-N
103534
CCC(C(=O)C)C(=O)OC
C7H12O3
MFCD01632136
methyl 2-ethylacetoacetate, butanoic acid, 2-ethyl-3-oxo-, methyl ester, methyl-2-ethylacetoacetate, 2-ethyl-3-oxobutanoic acid, methyl ester, methyl ethylacetoacetate, methyl acetylbutyrate, methyl 2-acetylbutyrate, acmc-1b06q, methyl 2-ethyl-3-oxobutyrate, methyl 2-ethyl-3-oxo-butyrate
methyl 2-ethyl-3-oxobutanoate

Specifications

51756-08-2
189°C to 191°C
1.423
methyl 2-ethylacetoacetate, butanoic acid, 2-ethyl-3-oxo-, methyl ester, methyl-2-ethylacetoacetate, 2-ethyl-3-oxobutanoic acid, methyl ester, methyl ethylacetoacetate, methyl acetylbutyrate, methyl 2-acetylbutyrate, acmc-1b06q, methyl 2-ethyl-3-oxobutyrate, methyl 2-ethyl-3-oxo-butyrate
CCC(C(=O)C)C(=O)OC
144.17
144.17
Methyl 2-ethylacetoacetate
1.01
C7H12O3
MFCD01632136
YXLVLOWNJCOOAU-UHFFFAOYSA-N
methyl 2-ethyl-3-oxobutanoate
103534
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 257-381-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications