Learn More
Brady™ MetaLabel™ B-428 Metallized Polyester BMP71 Labels
For use in commercial/industrial applications. Brady B-428 metallized polyester labels with permanent acrylic adhesive are for use with BMP71 printer.
Brand:  Brady™ 114953
 View more versions of this product
Product Code. 11747124
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Specifications
UL Recognized, CSA Accepted, Halogen Free (DIN VDE 0472/part 815)
Acrylic, Permanent
Matte
Asset Tracking Label
BMP™71 Printer
100/Pk.
3 x 1 in. (W x H)
-70°C to +120°C
0.112 mm
76.20 x 25.40 mm
76.20 mm
Chemical, Corrosion, Humidity, UV and Weather-Resistant
Light Gray
Metallized Polyester (B-428)
Asset tracking, Rating plates, General purpose ID
1
Rectangle
76.2 x 25.4 mm (W x H)
0.0044 in.
79.38 mm
25.40 mm