missing translation for 'onlineSavingsMsg'
Learn More

Bromodichloromethane in Methanol 100μg/mL, Fisher Chemical™

Product Code. 12979673
Click to view available options
Quantity:
1 mL
Packaging:
Ampoule(s)
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Concentration 100μg/mL
CAS 75-27-4
Packaging Ampoule(s)
Quantity 1 mL
Molecular Formula CHBrCl2
UN Number 1230
InChI Key FMWLUWPQPKEARP-UHFFFAOYSA-N
SMILES C(Cl)(Cl)Br
IUPAC Name bromo(dichloro)methane
Molecular Weight (g/mol) 163.823
PubChem CID 6359
ChEBI CHEBI:34591
Solvent or Matrix Methanol
Chemical Name or Material Bromodichloromethane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Standard solution Bromodichloromethane 100ug/ml in methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'