missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercury(Ii) Oxide Yellow, ≥99.0%, ACS Reagent, Honeywell™ Fluka™
ACS Reagent,≥99.0%
£32.67 - £309.09
Chemical Identifiers
CAS | 21908-53-2 |
---|---|
Molecular Formula | HgO |
Molecular Weight (g/mol) | 216.59 |
MDL Number | MFCD00011045 |
InChI Key | UKWHYYKOEPRTIC-UHFFFAOYSA-N |
Synonym | mercuric oxide, mercury ii oxide, mercury oxide, mercuric oxide, red, red mercuric oxide, mercuric oxide, yellow, santar, red precipitate, mercury monoxide |
IUPAC Name | oxomercury |
SMILES | O=[Hg] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15682080
|
Honeywell-Fluka
221082-5G |
5 g | Glass Bottle |
£32.67
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15662080
|
Honeywell-Fluka
221082-100G |
100 g | Glass Bottle | N/A | |||||
15672080
|
Honeywell-Fluka
221082-500G |
500 g | Glass Bottle |
£309.09
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
21908-53-2 | |
216.59 | |
UKWHYYKOEPRTIC-UHFFFAOYSA-N | |
oxomercury |
HgO | |
MFCD00011045 | |
mercuric oxide, mercury ii oxide, mercury oxide, mercuric oxide, red, red mercuric oxide, mercuric oxide, yellow, santar, red precipitate, mercury monoxide | |
O=[Hg] |
Specifications
21908-53-2 | |
HgO | |
UN1641 | |
UKWHYYKOEPRTIC-UHFFFAOYSA-N | |
oxomercury | |
216.59g/mol | |
ACS Reagent |
HgO | |
MFCD00011045 | |
mercuric oxide, mercury ii oxide, mercury oxide, mercuric oxide, red, red mercuric oxide, mercuric oxide, yellow, santar, red precipitate, mercury monoxide | |
O=[Hg] | |
216.59 | |
≥99.0% | |
Mercury(Ii) Oxide Yellow |
Safety and Handling
Missing Phrase - N15.00950417-P260-P262-P280-P302 + P352 + P310-P304 + P340 + P310
H300 + H310 + H330-H373-H410
missing translation for 'dotInformation' : 6.1/PGII
missing translation for 'einecsNumber' : 244-654-7
missing translation for 'rtecsNumber' : OW8750000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Mercury(II) oxide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Mercury monoxide
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title