missing translation for 'onlineSavingsMsg'
Learn More

Mercury(Ii) Oxide Red, ACS Reagent, ≥99.0%, Honeywell™ Fluka™

Product Code. 15611970 Shop All Honeywell Fluka Products
5 g, Glass Bottle
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21908-53-2
Molecular Formula HgO
Molecular Weight (g/mol) 216.59
MDL Number MFCD00011045
InChI Key UKWHYYKOEPRTIC-UHFFFAOYSA-N
Synonym mercuric oxide, mercury ii oxide, mercury oxide, mercuric oxide, red, red mercuric oxide, mercuric oxide, yellow, santar, red precipitate, mercury monoxide
IUPAC Name oxomercury
SMILES O=[Hg]

Specifications

Melting Point >400°C
Quantity 5 g
Linear Formula HgO
UN Number UN1641
Formula Weight 216.59g/mol
Percent Purity ≥99.0%
Grade ACS Reagent
Packaging Glass Bottle
Appearance Powder
Chemical Name or Material Mercury(Ii) Oxide Red
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mercury(Ii) Oxide Red, ACS Reagent, ≥99.0%, Honeywell™ Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.