Learn More
MED8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555829
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: VIIPLVLSPD RDEDLMRQTE GRVPVFSHEV VPDHLRTKPD PEVEEQEKQL TTDAARIGAD AAQKQ Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 32%.
This gene encodes a protein that is one of more than 20 subunits of the mediator complex, first identified in S. cerevisiae, that is required for activation of transcription. The product of this gene also interacts with elongins B and C, and CUL2 and RBX1, to reconstitute a ubiquitin ligase. Two alternative transcripts encoding different isoforms have been described.Specifications
MED8 | |
Polyclonal | |
Unconjugated | |
Med8 | |
2210021A15Rik; AB041805; Activator-recruited cofactor 32 kDa component; ARC32; Med8; Mediator complex subunit 8; mediator of RNA polymerase II transcription subunit 8; mediator of RNA polymerase II transcription subunit MED8; mediator of RNA polymerase II transcription, subunit 8 homolog; MGC17544; MGC19641; MNCb-2386; zgc:76877; zgc:76877 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
112950, 362575, 80509 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96G25, Q9D7W5 | |
Med8 | |
Recombinant protein corresponding to Human MED8 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only