missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCT2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552225
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: GPNQTTSKSK NKTGKTEDDS SPKKIKTKKS TWEKVNKYLD FS Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 45%.
Tissues with few or no mitochondria, such as erythrocytes and tumor cells, depend largely on glycolysis to generate ATP. The major end products of glycolysis, pyruvate and lactate, must be eliminated from these cells to enable continued glycolytic flux and prevent toxic effects. H+/monocarboxylate transporters mediate the transport of lactate and pyruvate. Human MCT2 has a high affinity for the transport of pyruvate.Specifications
MCT2 | |
Polyclonal | |
Unconjugated | |
SLC16A7 | |
4921534N07Rik; MCT 2; Mct2; monocarboxylate transporter; Monocarboxylate transporter 2; Slc16a7; solute carrier family 16 (monocarboxylate transporter), member 7; solute carrier family 16 (monocarboxylic acid transporters), member 7; solute carrier family 16 member 7; solute carrier family 16, member 7 (monocarboxylic acid transporter 2) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9194 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60669 | |
SLC16A7 | |
Recombinant protein corresponding to Human MCT2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only