missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mCherry Monoclonal Antibody (16D7), Alexa Fluor™ 594, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen M11240
Code : NEW
Additional Details : Weight : 0.25000kg
Description
MCherry is a fluorophore (a fluorescent molecule) used in biotechnology as a tracer to follow the flow of fluids, as a marker when tagged to molecules and cells components. mCherry is a monomeric fluorescent construct which absorbs at 587 nm and emits at 610 nm. It is resistant to photobleaching and is stable. It matures quickly, with a t 0. 5 of 15 minutes, allowing it to be visualized soon after translation.Specifications
mCherry | |
Monoclonal | |
Alexa Fluor 594 | |
mCherry | |
Full-length mCherry was used as the immunogen | |
50 Tests | |
Primary | |
Antibody | |
IgG2a |
Flow Cytometry | |
16D7 | |
PBS with 5mM sodium azide; pH 7.5 | |
Rat | |
Affinity chromatography | |
RUO | |
4° C | |
Liquid | |
Tag |