missing translation for 'onlineSavingsMsg'
Learn More

MBL™ Type 2 Class B Graduated Pipettes

Manufactured from soda-lime glass with robust tooled jets

Brand:  MBL PRA804B

Code : 33

Additional Details : Weight : 0.19000kg

 View more versions of this product

Product Code. 12560046

  • £19.10 / Pack of 5
Estimated Shipment: 29-05-2024
to see stock.
Add to basket

Description

Description

  • Calibrated to deliver from any graduation line down to zero at jet
  • Graduations in permanent amber stain which is fused into glass to ensure highest resistance to aggressive washing solutions
Specifications

Specifications

ISO 835
Yes
5/Pk.
5 mL
Soda Lime Glass
0.05 mL
±0.05 mL
Pipette
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers