Learn More
Thermo Scientific™ Maximum Recovery Diluent
Achieve maximum recovery of microorganisms during dilution with this protective, isotonic diluent.
Brand:  Thermo Scientific™ BO0348D
Code : 64
Additional Details : Weight : 0.80010kg
 View more versions of this product
Product Code. 12756949
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Achieve maximum recovery of microorganisms during dilution with this protective, isotonic Thermo Scientific™ Oxoid™ Maximum Recovery Diluent. 
The presence of peptone in the diluent provides viable counts to ensure accurate quantitative procedures are performed. The isotonic strength of the diluent ensures recovery of organisms from various sources which may be vulnerable in distilled water or aqueous suspensions. The low peptone concentration in the diluent reduces organism multiplication in the sample for at least 45 minutes during the dilution stage1,2
Maintain the viability of organisms during dilution process with Maximum Recovery Diluent with low levels of peptone.
  • Allows accurate quantitative procedures to be performed with minimal reduction in viable count.
  • Reduces organism multiplication in the sample for at least one hour during the dilution stage.
  • Ready-to-use.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
Colorless, clear
Maximum Recovery Diluent
United Kingdom
9.0g + 0.3g
24/Pk.
2 to 25°C
Universal bottles
Agar
Twenty four universal bottles with screw cap closures in a box
Product Suggestions
Product Suggestions
Videos
SDS Documents