Learn More
Thermo Scientific™ Maxima H Minus First Strand cDNA Synthesis Kits Promotion
Synthesize long cDNAs up to 20 kb at elevated temperatures (up to 65°C) with this complete system for highly efficient synthesis of first strand cDNA
Brand: Thermo Scientific™ K1652
This item is not returnable.
View return policy
Description
- Increased reaction temperatures -the first strand of cDNA can be synthesized within the 42 to 65°C temperature range
- High yields of full-length first strand cDNA -with RNA templates up to 20 kb
- Flexible priming -oligo(dT)18, random hexamer, or gene-specific primers
Specifications
RT-PCR | |
100 reactions |