Learn More
Fisherbrand™ Max Mono II Monocular Biological Microscope
Ergonomically designed for everyday use with easily accessible controls for stage movement coarse and fine focus plus varying light intensity.
Brand:  Fisherbrand™ 1201.4000M
Product Code. 12914361
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Reversed 4-position turret for easy access to stage
  • With carrying handles
  • Full mechanical stage
  • Pre-centered Abbe condenser
  • With three achromatic objectives – 4x, 10x, 60x
  • 20W illumination and comes with a blue filter and dust cover
Specifications
Specifications
Abbe
Rotating
Max Mono II
220/240 V 50/60 Hz
50/60 Hz
10X/18 mm
Halogen
Achromatic
220/240 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products