missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Malonaldehyde bis(phenylimine) monohydrochloride, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Malonaldehyde bis(phenylimine) monohydrochloride is a useful fluorescent cyanine dye.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Malonaldehyde bis(phenylimine) monohydrochloride is a useful fluorescent cyanine dye.
Solubility
Soluble in water (partly).
Notes
Hygroscopic, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Malonaldehyde bis(phenylimine) monohydrochloride is a useful fluorescent cyanine dye.
Solubility
Soluble in water (partly).
Notes
Hygroscopic, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Melting Point | ∽215°C (decomposition) |
Quantity | 25 g |
Solubility Information | Soluble in water (partly). |
Formula Weight | 258.75 |
Percent Purity | 98% |
Chemical Name or Material | Malonaldehyde bis(phenylimine) monohydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'