missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Oxide, Powder, U.S.P. - F.C.C., J.T. Baker™
£287.00 - £456.00
Chemical Identifiers
CAS | 1309-48-4 |
---|---|
Molecular Formula | MgO |
Molecular Weight (g/mol) | 40.30 |
MDL Number | MFCD00011109 |
InChI Key | CPLXHLVBOLITMK-UHFFFAOYSA-N |
Synonym | magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag |
PubChem CID | 14792 |
IUPAC Name | oxomagnesium |
SMILES | O=[Mg] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15508014
|
Avantor J.T.Baker
2480-01 |
500 g | Poly Bottle |
£287.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15518014
|
Avantor J.T.Baker
2480-05 |
2 kg | Poly Pail | N/A | |||||
Chemical Identifiers
1309-48-4 | |
40.30 | |
CPLXHLVBOLITMK-UHFFFAOYSA-N | |
14792 | |
O=[Mg] |
MgO | |
MFCD00011109 | |
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag | |
oxomagnesium |
Specifications
1309-48-4 | |
MgO | |
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag | |
O=[Mg] | |
40.30 | |
40.3 | |
1L = 3.58kg |
Powder | |
MFCD00011109 | |
CPLXHLVBOLITMK-UHFFFAOYSA-N | |
oxomagnesium | |
14792 | |
FCC/USP | |
Magnesium Oxide |