missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAFF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12771-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a basic leucine zipper (bZIP) transcription factor that lacks a transactivation domain. It is known to bind the US-2 DNA element in the promoter of the oxytocin receptor (OTR) gene and most likely heterodimerizes with other leucine zipper-containing proteins to enhance expression of the OTR gene during term pregcy. The encoded protein can also form homodimers, and since it lacks a transactivation domain, the homodimer may act as a repressor of transcription. This gene may also be involved in the cellular stress response. Multiple transcript variants encoding two different isoforms have been found for this gene.Specifications
MAFF | |
Polyclonal | |
Unconjugated | |
MAFF | |
hMafF, MAFF, Transcription factor MafF, U MAF | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17133, 23764, 366960 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.42 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O54791, Q9ULX9 | |
Maff | |
MAFF Fusion Protein Ag3460 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MAFF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction