Learn More
IBA Lifesciences M6 Adapter Set
Adapters for H-PR cartridges
Brand: IBA Lifesciences 2-1022-001
This item is not returnable.
View return policy
Description
TheStrep-tag™ purification system is based on the highly selective and easily controllable interaction between theStrep-tag™II peptide and the biotin binding site of a specially engineered streptavidin calledStrep-Tactin™.Strep-tag™II bindsStrep-Tactin™ nearly 100 times tighter than streptavidin, but elutes under gentle, physiological conditions.- Rapid one-step purification under physiological conditions
- Unsurpassed purity and bioactivity
Specifications
M6 Adapter Set | |
It Contains 2 x Female Peek M6 to 10-32 Male | |
For H-PR cartridges |
1 Ea. | |
Column Accesory |