missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LZIC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14543-1-AP-150UL
This item is not returnable.
View return policy
Description
LZIC belongs to the CTNNBIP1 family. It is ubiquitously expressed with highest levels in kidney. LZIC is up-regulated in several cases of gastric cancers.Specifications
LZIC | |
Polyclonal | |
Unconjugated | |
LZIC | |
LZIC, Protein LZIC | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
366507, 69151, 84328 | |
-20°C | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5PQN7, Q8K3C3, Q8WZA0 | |
Lzic | |
LZIC Fusion Protein Ag6047 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |