missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Lysozyme from Chicken Eggwhite Type VI
3X crystallized, salt-free, albumin-free
POA
Specifications
Concentration | 3X |
---|---|
Color | Off-White |
pH | 11 |
For Use With (Application) | Lysing E. coli and Streptomycetes for extraction purposes such as extracting group specific Antigen |
Content And Storage | 0°C |
Description
Activity: ≥23,500 Shugar units/mg. Unit Definition: One unit will produce a decrease in A450 of 0.001 per minute at pH 6.24 and 25°C using Micrococcus lysodeikticus as substrate. Lyophilized.
Specifications
3X | |
11 | |
0°C | |
Lysozyme | |
Surface-active reagents such as Dodecyl Sulfate, Alcohols and Fatty Acids, Imidazole and Indole derivatives are inhibitors via formation of change-transfer complexes |
Off-White | |
Lysing E. coli and Streptomycetes for extraction purposes such as extracting group specific Antigen | |
Chicken egg white | |
14388 | |
Lyophilized |